Would you like to log in again?
Chemware® Teflon® Resin PFA Jars, Saint-Gobain
Supplier: Saint-Gobain
Chemware®
D1069030 D1069027 D1069025 D1069706 D1069029 D1069024 D1069031
36324-002EA 281.58 USD InStock
36324-002 36324-001 36324-000 36324-006 36324-005 36324-004 36324-003
Chemware® Teflon® Resin PFA Jars, Saint-Gobain
Bottles Plastic Bottles
Store solutions, pastes, and bulk materials without leaching, corrosion, contamination, or evaporation.
  • Knurled top for sure grip. Large mouth for easy access
  • Secondary inner seal for safe storage of any substance
  • Heat to 260 °C (500 °F). Freeze to –195 °C (–320 °F)
  • Stackable, easily cleaned
Certifications: Jar 36324-007 meets EPA requirement for soil sampling.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty