Would you like to log in again?
Nalgene® Polycarbonate Multipurpose Jars with Covers, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
DS5300-0507 DS5300-9609 DS5300-9910
36325-360EA 329.28 USD InStock
36325-360 36324-989 36325-008
Nalgene® Polycarbonate Multipurpose Jars with Covers, Thermo Scientific
Jars General Use Jars
Nalgene® Polycarbonate Multipurpose Jars are useful as water baths, aquaria, terraria or freeze-drying chambers
Cat. Nos. DS5300-0504; DS5300-0507 may be used for vacuum service if not autoclaved. For vacuum service, see Cat. Nos. 5305 and DS5320.
You can drill, tap or apply clamps to attach heating or cooling equipment. The covers permit the jars to be stacked.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty