Would you like to log in again?
Nalgene® High-Density Polyethylene Jerricans, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2240-0015 2240-0050 2243-0013 2243-9013 2240-0025
36382-022CS 524.26 USD InStock
36382-022 36382-066 73520-057 36382-044 73520-059
Nalgene® High-Density Polyethylene Jerricans, Thermo Scientific
Bottles Jerricans
Sturdy, leakproof containers are intended for hard use.
A wide stance and low center of gravity provide greater stability. Integral spout is long enough for accurate pouring, and features a strap-fastened cap. Molded-in graduations are provided in liters and gallons. Recessed bottom provides second handgrip for pouring. Jerricans 73520-057 and -059 weigh 13 kg (28.7 lbs.) at their normal fill volume, just right for unassisted lifting. Jerrican 73520-059 features a second spout with a tethered cap for vented pouring. This provides access for refilling when the 53B spout is fitted with connections to equipment.
Ordering information: Jerricans are supplied with 53B (buttress-style) polypropylene closures. Jerrican 73520-059 also comes with a 38-430 polypropylene closure.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty