Would you like to log in again?
Nalgene® HDPE Heavy-Duty Wide Mouth Jug, Thermo Scientific
Supplier: Thermo Scientific
2241-0050
36382-075CS 766.08 USD InStock
36382-075
Nalgene® HDPE Heavy-Duty Wide Mouth Jug, Thermo Scientific
Bottles Plastic Bottles
Nalgene heavy-duty wide mouth jug feature the largest opening, 120 mm (4") in the Nalgene carboy line.
The jugs come with a molded-in handle and recessed bottom which allows for easier filling and emptying without spills. The jug features a wide stance and a low center of gravity which assures stability. Also the jug contains a large neck for easy cleaning, and a space saving rectangular shape which permits efficient storage. Graduations are molded into the gallons and liters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty