Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PYREX® Resin Reaction Kettles with Covers, Corning

Supplier: Corning

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PYREX®
6947-2LCO 6947-3L 6947-4LBO 6947-4L 6947-2LBO 6947-500BO 6947-2L 6947-3LBO 6947-1LBO 6947-500CO 6947-1L 6947-500
36390-086CS 1320.85 USD InStock
36390-086 89091-294 36390-064 36390-042 36390-020 89091-648 36390-100 89091-208 89091-650 89091-422 89091-354 89091-388
PYREX® Resin Reaction Kettles with Covers, Corning
Reaction Vessels Reaction Vessels, Cylindrical Shape
PYREX® borosilicate glass.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

These vessels have lids with openings to accommodate four Standard Taper joints. Interchangeable Standard Taper joints allow quick assembly with condensers, stirrers and funnels.

Flanges of lids and vessels are finely ground for a tight seal.

For use with heating mantle only.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty