Would you like to log in again?
SP Bel-Art Protective Laboratory Labeling System, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F13450-0000
36440-107EA 132.38 USD InStock
36440-107
SP Bel-Art Protective Laboratory Labeling System, Bel-Art Products, a part of SP
Label and Tape Dispensers
The protective labeling system provides labels that stand up to harsh laboratory conditions.
  • Durable dispenser
  • Clear polypropylene tape
  • High resistance to chemicals and moisture
  • Tape will withstand freezer, autoclave, alkalis, and organic solvents
Durable tape dispenser is molded of strong ABS plastic with a steel serrated cutting blade.
Test results prove that it remains securely affixed with no discoloration after being autoclaved for 40 minutes at 125 °C (258 °F), as well as after one week in liquid nitrogen at –196 °C (–321 °F). All tape sizes are 1mil (0.001") thick with 1mil (0.001") of acrylic adhesive.
White paper labels accept any kind of marker and are perfect for use under the clear tape to label your labware, bottles, or samples. The tape will protect them from the freezer, autoclave, acids, alkalis, and organic solvents.
A tape roll is held in place by an aluminum roller and spring-loaded cover.
Ordering information: All items are sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty