Would you like to log in again?
Dipper, PTFE, Dynalon
Supplier: Dynalon
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
307024-0500
36495-114EA 466.73 USD InStock
36495-114
Dipper, PTFE, Dynalon
Samplers Sample Dippers
Totally inert and non-contaminating
Extendible screw-in shafts have a steel core for rigidity and are fully encapsulated in PTFE. The basic unit consists of a beaker with a single 24" (600mm) shaft.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty