Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

UVP Compact and Handheld Lamps, Analytik Jena

Supplier: Analytik Jena
95-0021-12 95-0018-02 95-0005-05 95-0019-06 95-0019-04 95-0104-01 95-0004-09 95-0342-02 95-0164-02 95-0343-02 95-0004-10 95-0007-05 95-0130-01 95-0016-15 95-0343-01 95-0017-09 95-0017-10 95-0019-03 95-0016-14 95-0017-09 95-0021-12 95-0342-01 95-0341-01 95-0005-06 95-0341-02 95-0104-02 95-0006-03 95-0006-02 95-0164-01 95-0018-02 95-0007-06 95-0019-14 95-0016-14
89237-394EA 1451.88 USD InStock
89237-394 74670-010 89237-392 36582-158 CA36582-045 74670-012 89237-390 21474-616 CA36580-058 89237-358 36585-065 74690-002 89237-356 21474-622 21474-620 89237-340 89237-384 36582-045 21475-474 36585-777 74670-024 74670-002 36585-733 36580-058 36585-815 CA36585-065 74670-004 89237-388 89237-386 470204-312 36582-040 21474-634 36580-172
UVP Compact and Handheld Lamps, Analytik Jena
Lights UV Lights, Lamps and Bulbs
UVP Compact and Handheld Ultraviolet Lamps use UV wavelengths which are an essential multipurpose tools for laboratory applications. The lamps are lightweight and ergonomically designed for handheld use.

Longwave (365 nm) can be used for bacterial identification, inspection, specimen staining and titration processes, gel electrophoresis, thin layer chromatography, visualization of DNA and RNA and fluorescence of other materials are among the applications

UVP Compact Lamps feature 4 W intensity format and the UVP Hanndheld Lamps utilize a higher 6 W intensity.

For applications requiring all three UV wavelength the patented UVP 3UV™ Multi-Wavelength Lamp, houses multiple wavelengths: longwave, shortwave and midrange (254/302/365 nm UV) in one lamp for multiple applications.

UVP Fraud Detection Lamps use longwave ultraviolet light to check the authenticity of signatures, documents, tickets, passports, credit cards and currency. The lamp is a cost-effective solution to security problems. Invisible inks or altered documents will fluoresce for visibility. The lamp is used for instant document verification, detection of forged documents, currency and signatures.

Shortwave (254 nm) can be used for sterilization/germicidal, DNA/RNA fluorescence and analysis, photochemistry and more.

Certifications: CSA certified. CE compliant.

Accessories information: Additional accessories available:

Use the compact lamps with optional J-124 lamp stand for hands free operation. Handheld lamps can be used with the optional J-129 lamp stand for hands free operation.

Mount the lamps onto the optional C-10 Viewing Cabinet which provides a darkened environment for viewing or exposing samples with UV.

Ordering information: One year warranty.

Caution: Eye and face protection is essential for anyone working with UV sources which can cause burning to unprotected eyes and skin. Eyewear blocks wavelengths from 200 - 400 nm. Spectacles are made of impact resistant polycarbonate with top and side shield design. Goggles feature lightweight design and flexible rubber trim. Air vents allow air flow. Faceshield covers the entire face and neck area.

Goggles and faceshield comply with ANSI specifications for eye and face protection.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty