Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

UVP Blak-Ray® B-100 Series Lamps, Analytik Jena

Supplier: Analytik Jena
UVP Blak-Ray®
95-0044-03 95-0044-02 95-0044-22 95-0128-01
36595-001EA 1453.78 USD InStock
36595-001 36595-002 36595-025 21474-628
UVP Blak-Ray® B-100 Series Lamps, Analytik Jena
Lights UV Lights, Lamps and Bulbs
The UVP Blak-Ray B-100 Series lamps are available in a variety of styles: All "P" series lamps come with the special heat-resistant plastic Cool-TouchTM housing which allows users to handle the lamp head regardless of how long the lamp has been operating. These rugged lamps can be placed face down on a working surface without damage to the filter. The non "P" models are built with an aluminum lamp head.

  • Available in a variety of styles
  • Lightning: Overhead: White; Trans: UV

A 100 watt spot bulb is standard with all models (except the B-100SP); the bulb is rated at 5000 hours.

The lamp head connects to the transformer base for hands free use (all models except B-100SP) allowing the lamp head to rotate a full 360°; remove the lamp from the base for additional mobility.

B-100A and A-100AR–The longwave B-100A is manufactured with a scratch-resistant aluminum housing which connects to ballast base with an 8 foot cord; 8 foot secondary cord is included. The 100 watt longwave UV bulb is excellent for inspection applications. For increased mobility, select the B-100AR which includes 6.1 m (20 ft.) primary and secondary cords. Uses for these lamps include non-destructive testing and fluorescence inspection.

The B-100Y models features a yellow filter which blocks wavelengths shorter than 500 nm and produces three strong mercury lines at 543, 574 and 576 nm. The high contrast light is produced for fine surface particle detection. Particles down to 10 microns are routinely visible with this lamp. B-100Y is built with the aluminum housing; B-100YP with a plastic housing. These lamps are excellent for semiconductor wafer inspection as most photoresist absorption occurs below 450 nm.

Self-ballasted bulb included with the B-100SP has an average life of 450 to 2000 hours depending on use patterns.

Certifications: CSA certified. CE compliant.

Accessories information: Dimensions: 216D x 140 Dia. mm (8.5 x 5.5")

Ordering information: One-year warranty. Additional accessories available include Finger Guard–Prevents fingers from touching the bulb; Lamp Funnel–Directs the UV source to a specific location; Lamp Visor–Shields the user from the blue haze emitted from the lamp; Exposure Box; UV Blocking Eyewear; Reveal Leak Detection Additives. For more information, contact your Avantor representative.

Order Now
 

ORDER

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty