Would you like to log in again?
TouchNTuff® 69-318 Natural Rubber Latex Disposable Gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Conform®
6931800L 6931800S 693180XL 516701
36888-210CS 247.2 USD InStock
36888-210 36888-212 36888-214 36888-216
TouchNTuff® 69-318 Natural Rubber Latex Disposable Gloves, Ansell
Gloves
SType 69-318 gloves are 100% natural rubber latex.
  • Powder-free
  • Ambidextrous
  • Textured grip
Gloves are suitable for laboratory analysis/technical work, food processing and handling, quality control, electronics assembly, as well as light-duty maintenance and cleanup.
Certifications: Gloves qualify as Class I Medical Devices. Component materials comply with FDA regulations for food contact.
Caution: This product contains natural rubber latex which may cause allergic reactions in some individuals.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty