Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Measuring Magnifier, Bausch & Lomb®

Supplier: Bausch & Lomb

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

813435 813438 813436 813437 813439
36929-060EA 103.18 USD InStock
36929-060 36929-049 36929-005 36929-027 36927-000
Measuring Magnifier, Bausch & Lomb®
Magnifiers
For quick and accurate measuring, and for convenient inspection of small detail

General-purpose scale measures radii from 1/16 to 3/8" (angles in single degrees to 90°; 0.005" and 0.1mm scales; line widths of 0.001, 0.002, and 0.003"). Inch scale measures to 0.75" in 0.005" intervals. Metric scale measures to 20mm in 0.1mm intervals. Protractor scale measures 360° in single degrees.

Precision ground, 7x Hastings triple aplanatic lens can be quickly focused. Long transparent body utilizes maximum degree of light.

Ordering information: Leather case included. Please order magnifier scales separately.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty