Would you like to log in again?
VWR® Freezer Thermometer –100 °C, Platinum
Supplier: VWR
VWR®
90029-93
36934-160EA 139.35 USD InStock
36934-160
VWR® Freezer Thermometer –100 °C, Platinum
Thermometers Digital Thermometers
Thermometer accurately monitors temperatures in freezers, water baths, heating blocks, incubators, and refrigerators.
  • Traceable®
  • Does not affect seal of closed doors
  • Fast-response probe
  • Designed for use with liquids, air/gas, and frozen material
A 3m (10') ultra-thin ribbon micro-cable permits freezer doors to close on it without affecting the seal. The fast-response, 100Ω platinum probe is designed for use with liquids, air/gas, and frozen material.
Certifications: A serial-numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with adjustable probe holder, pre-cut Velcro® squares, magnetic strips, chemical-resistant carrying case, and 9V alkaline battery.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: VWRCATALYST™ provides convenient and efficient re-calibration services for your digital thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty