Would you like to log in again?
VWR® Traceable® Water-Resistant Food Thermometer
Supplier: VWR
VWR®
VC2068 93677-36 93677-40 VC1068 VC4420
36934-168EA 66.69 USD InStock
36934-168 10171-502 36934-166 10171-618 10171-330
VWR® Traceable® Water-Resistant Food Thermometer
Thermometers Digital Thermometers
Ideal for food labs, food processing, food preparation, and brewing.
  • Traceable®
  • Durable ABS plastic body
  • Flat-profile design
  • Water-resistant
  • Lightweight
Waterproof construction allows thermometer to be used in labs, plants, wash-down areas, and the field. Designed with membrane switches and ABS body, the unit eliminates dangers inherent in breakable glass and toxic mercury thermometers. Unique holder allows stainless steel probe to be positioned at any height while attached to beakers, stainless cylinders, or vats. Flat-profile design, light weight, and built-in pocket clip make it ideal to carry in a shirt pocket. Probe guard snaps to handle to extend overall length to 27.3 cm (103/4"). Readings are updated every second. Features a bright 6mm (1/4") LCD display. Operates continuously for 18 months on a single battery.
Certifications: A serial-numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with silver-oxide battery, keychain, and lanyard.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: VWRCATALYST™ provides convenient and efficient re-calibration services for your digital thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty