Would you like to log in again?
VWR® Traceable® Digital Count-Up Controllers
Supplier: Avantor
VWR® Traceable®
93678-79
36934-170EA 238.3 USD InStock
36934-170
VWR® Traceable® Digital Count-Up Controllers
Timers Time Switches
Controllers perform reproducible stirring, heating, or pumping times.
  • Internal battery maintains programs in event of a power outage
  • Used for timed shutdown or repeated on/off timing during any test
  • LED display
Can be used for timed shutdown or repeated on/off timing during any test.
The turn-off controller counts up and turns off equipment at the end of a set period. Turn-off controller is programmable in three interval settings: 0.01 to 99.99 seconds, 1 to 99 minutes, or 1 to 99 hours.
Manual override pause switch instantly stops equipment without disturbing program. Intuitive operation allows programming in less than five seconds. Internal NiCad battery maintains programs in the event of a power outage. Three-pronged grounded receptacle relay controls apparatus to 120 V, 15 A, 1800 W, and ¹/₃ hp. Red LED indicates the on/off status of attached equipment. LCD digits are 1.3 cm (¹/₂") high.
Certifications: A serial-numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate traceability to standards provided by the National Institute of Standards and Technology.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty