Would you like to log in again?
VWR® Traceable® Infrared Thermometer with Type-K Probe
Supplier: VWR
VWR®
90012-56
36934-178EA 244.3 USD InStock
36934-178
VWR® Traceable® Infrared Thermometer with Type-K Probe
Thermometers Digital Thermometers
Non-invasive, no-touch measurements are ideal for food preparation, life sciences, pharmaceuticals, petroleum products, cleanrooms, electronics, and field use.
  • Traceable®
  • Non-invasive measurements
  • Memory functions recalls past temperature readings
  • Backlit display for low-light conditions
  • Hold freezes the display at the current reading
Simple operation allows unit to take readings in less than a second
The field of view is 11:1; thermometer can read a spot of 5 cm (2") from 55.9 cm (22") away. Emissivity is adjustable from 0.1–1.00 in 0.01 steps.
Unit takes Celsius and Fahrenheit readings of solids, semi-solids, and liquids. Minimum/maximum feature allows viewing of highest and lowest readings at any time. Thermometer's memory recalls individually captured temperatures and displays the average of all readings made and the difference between two readings. Audible, user-settable, high and low alarms signal out-of-range conditions. Backlighting permits viewing in low-light conditions. Display indicates low battery.
Certifications: A serial-numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with Type-K probe and AAA alkaline batteries. Supplied probe does not cover full temperature range of thermometer. Additional Type-K probes with various temperature ranges are available separately; contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty