Would you like to log in again?
OptiVISOR® Binocular Headband Magnifier
Supplier: Excelta
OptiVISOR®
DA-7 BLUE
36968-007EA 149.46 USD InStock
36968-007
OptiVISOR® Binocular Headband Magnifier
Magnifiers
For close or precision work requiring visual accuracy
Continuous headband with dial adjustment for comfortable wear. Visor and lens pivot up and out of the way for normal vision when not in use. Fits over regular glasses. Magnifies with 15.2cm (6") focal length.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty