Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

KIMAX® Capillary Melting Point Tubes, Kimble Chase,DWK Life Sciences

Supplier: DWK Life Sciences
KIMAX®
34505-99 34502-99 34500-99 34507-99
36982-008CS 373.35 USD InStock
36982-008 36983-000 36984-003 36984-025
KIMAX® Capillary Melting Point Tubes, Kimble Chase,DWK Life Sciences
Tubes Melting Point Tubes and Capillaries
Capillary tubes available in 100 mm length with both ends open, or 90 mm length with one end sealed.

Tubes 36982-008 and 36984-003 have an O.D. of approx. 1.5 to 1.8 mm with a 0.2 mm wall thickness.

Tubes 36983-000 and 36984-025 comply with requirements given in the United States Pharmacopoeia (USP) and have an I.D. of approx. 0.8 to 1.1 mm with a 0.25 mm wall thickness.

Delivery information: Packed in vials of 100 tubes, 20 vials per case.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty