Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Oxidation Pressure Vessel and Oxidata™ Software, Koehler
Supplier: Koehler Instrument
K10500
37000-034EA 6950.52 USD InStock
37000-034
Oxidation Pressure Vessel and Oxidata™ Software, Koehler
Petroleum Testing Apparatus
Precision machined stainless steel pressure vessel includes threaded body, lid, stem with filler rod and mounting flange, and needle valve for purging, pressurizing, and exhausting pressure vessel with oxygen
Available Oxidata™ pressure measurement systems (sold separately) run on Windows® XP and offer advanced, real time data acquisition and management for fuel and lubricant ASTM oxidation tests.
Oxidation pressure vessel conforms to specifications: ASTM D525.
Burst disk assembly ruptures at 223psi (1537kPa) to prevent unsafe pressure buildup. Vessel and stem interiors have a high polish to facilitate cleaning and prevent corrosion. Octagonal sections on the vessel and lid permit tight closures with a wrench.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty