Would you like to log in again?
Refrigerated Constant Temperature Kinematic Viscosity Baths, 220-240 V, Koehler
Supplier: Koehler Instrument
K22754
37000-150EA 36269.71 USD InStock
37000-150
Refrigerated Constant Temperature Kinematic Viscosity Baths, 220-240 V, Koehler
Baths Viscometer Baths
Measure flow under gravity or vacuum at precisely controlled temperatures.
  • 220-240 V model, not for use in USA
Insulated steel cabinet is finished in polyester epoxy and mounted on adjustable leveling feet. Chemical-resistant top surface has four round ports for viscometer holders and one port for thermometer holder. Built-in overtemperature protection is complemented by an integrated liquid sensor, which halts power to the heaters if liquid falls below the proper level.
Units feature microprocessor PID design and two decimal calibration offset. Simple push-button controls and dual digital displays permit easy setting and monitoring of bath temperature. Baths accommodate four glass capillary viscometers using 51 mm (2") round holders. Bath medium is contained in a clear dewar flask, and glare-free fluorescent backlighting provides high visibility through distortion-free viewing window. Air-cooled hermetic compressors provide efficient operation with the use of CFC-free refrigerants.
Certifications: Conform to specifications: ASTM D445, D2532, D6074, D6158; IP 71; ISO 3104; DIN 51550; FTM 791-305.
Ordering information: Supplied with a thermometer holder and four plastic viscometer port covers with handles.
Caution: 220-240 V model, not for use in USA. Corresponding model with voltage suitable for use in US (37000-944) is also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty