Would you like to log in again?
Sample Thieves (Bacon Bombs), Koehler
Supplier: Köhler Technische Produkte
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
K27795 K27761 K27701 K27782 K27760 K27770 K27785 K27772 K27771 K27762 K27792 K27781 K27791
37000-188EA 2796.04 USD InStock
37000-188 37000-210 37000-200 37000-212 37000-202 37000-214 37000-204 37000-206 37000-208 37000-192 37000-194 37000-196 37000-198
Sample Thieves (Bacon Bombs), Koehler
Samplers Fluid Samplers
Devices obtain samples from any level in storage tanks, tank cars, and drums
A plunger assembly opens to admit the sample when the thief strikes the bottom of the tank or is triggered by a secondary trip line. Plunger locking cam provides a tight closure during transport, except for 2.8cm (11/8") diameter models, which are designed for sampling through narrow pipes and openings.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty