Would you like to log in again?
VWR® Traceable® Food/Piercing Thermometer
Supplier: VWR
VWR®
93677-04 93676-99
77776-718EA 59.38 USD InStock
77776-718 37000-414
VWR® Traceable® Food/Piercing Thermometer
Thermometers Digital Thermometers
Measure the internal temperature of stored, frozen, or prepared food with the use of a piercing thermometer.
  • Traceable®
  • Shockproof
  • Read in °C or °F
  • Waterproof
Feature piercing stainless steel probes ideal for use in soil, brewing, food processing, refrigeration, and pharmaceutical applications. Waterproof construction. Shockproof from 1.5m (5'). LCD display measures 6.3mm (1/4") high. Reading is updated every second.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty