Would you like to log in again?
Sterile Specimen Cups, Covidien
Supplier: Covidien
8889207034 8889207026-
37000-632CS 260.83 USD InStock
37000-632 13915-772
Sterile Specimen Cups, Covidien
Sample Containers General Purpose Containers
Full turn screw thread provides an excellent leak-resistant seal
Translucent polypropylene container is tough, durable, non-breakable, and inert. It tolerates strong acids, bases, and most organic solvents. Sterile containers have a integrity seal or O.R. package. Cups are graduated in both a U.S. and metric scale. Individually wrapped and supplied with green cap. Meets the NCCLS Specimen Handling Guidelines for primary containers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty