Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
WypAll® X60 Wipers, Kimberly-Clark Professional®
Supplier: Kimberly-Clark
WYPALL®
34965 35421 35401 34790 34865 34965 34900 34955 34865 35401 34900 54015 35411 34790 35411 41083 35431 35421 34770
37002-020CS 358.15 USD InStock
37002-020 CA21903-022 21903-022 76326-016 37002-008 CA37002-006 CA37002-008 21903-020 CA37002-024 37002-004 CA37002-046 21903-040 CA37002-004 37002-006 37002-010 CA37002-020 37002-000 37002-046 37002-024
WypAll® X60 Wipers, Kimberly-Clark Professional®
Wipes Reusable Cleaning Cloths Cloths for General Applications
WypAll® X60 Cloths are absorbent, lightweight, and reinforced to stay strong in light-duty cleaning tasks.
  • Embossed surface traps and holds dirt and liquids
  • Can absorb more than 2× as much water as rags
  • Can be used in a range of medium- to heavy-duty applications
  • Contains no adhesives or binders, yet they hold together and don’t leave residues behind
  • Tough enough for big jobs, but soft enough for use on face and hands
  • Can be used wet or dry
Durable, uniformly reinforced towels for extra scrubbing and cleaning power. Embossed surface traps and holds dirt and liquids. Can be used in a full range of applications including utility companies, plant and fleet maintenance, machine shops, ore mines, and grocery stores. No adhesives or binders; wipers leave nothing behind. Tough enough for big jobs, yet soft enough for face and hands. Available in a variety of sizes and formats. Wipers 21903-040 and 37002-024 are hygienic washcloths that are excellent for both body cleansing and drying.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty