Would you like to log in again?
KIMTECH PREP® KIMTEX™ Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Kimtech™ Prep® Kimtx®
33570 33560 33570 33352 33241 33241 33560 33352
37002-050CS 225.2 USD InStock
37002-050 CA21908-646 CA21908-668 89000-690 37002-058 CA37002-058 37002-022 CA37002-022
KIMTECH PREP® KIMTEX™ Wipers, KIMBERLY-CLARK PROFESSIONAL®
Wipes Reusable Cleaning Cloths Cloths for General Applications
KIMTECH PREP® KIMTEX™ Wipers feature 100% polypropylene construction for cloth-like feel and durability.
Wipers provide a virtually lint-free cleaning performance and absorb quickly for big cleanups. Engineered for critical applications involving solvents, inks, and paints, they are also perfect for use with lubricants, oil, and grease. Acid-, base-, and solvent-resistant. Available in a variety of formats to meet every need. Color: blue. Applications include furniture finishing, engine assembly/repair, prepackaging wipe down, LCD panel assembly, appliance assembly, printing, and glass or window cleaning.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty