Would you like to log in again?
WYPALL® L20 Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WYPALL®
34607 47011 47758 47000 34607 47022 47022 47044 47758 47044
37002-030CS 157.8 USD InStock
CA89004-426 37002-030 CA37002-012 37002-048 14236-036 CA37002-048 37002-032 37002-012 CA37002-032 89004-426
WYPALL® L20 Wipers, KIMBERLY-CLARK PROFESSIONAL®
Wipes Disposable Wipes and Towels Wipes for General Applications
Paper wipers are excellent for large spills.
  • High-capacity single-use paper wipes
  • Textures side for better cleaning power
  • Contain no binder or glues
Perfect for parts and tool cleanups, mining operations maintenance, hydraulic leak absorption and wiping, general-purpose field utility wiping, conveyor and roller cleaning, and retail store maintenance and cleaning.
Packaging: Quarter-fold wipers are wrapped in stacks of 68. The BRAG™ box contains 176 wipers; Pop-Up™ boxes of 37002-048 contain 88 wipers.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty