Would you like to log in again?
Weller® Pyropen® Jr Self-Igniting Cordless Soldering Iron, Cooper Tools, Apex
Supplier: Apex
Weller® Pyropen®
WSTA6
37691-018EA 292.66 USD InStock
37691-018
Weller® Pyropen® Jr Self-Igniting Cordless Soldering Iron, Cooper Tools, Apex
Soldering Supplies Soldering Irons
Features self-igniting finger switch so no spark lighter is required
Iron heats up in 30 seconds. Adjustable power from 15 to 60 watts allows temperature control. Butane gas refill nozzle refills in 20 seconds. Knurl release switches from soldering to hot air in seconds. Handle can hold approximately 1 hour of butane gas reserve. With protective cap.
Ordering information: Packaged on a trilingual card.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty