Would you like to log in again?
Erem® Cutting Swiss Tweezers, Cooper Tools
Supplier: Cooper Tools
Erem®
15AGW
37691-628EA 397.3 USD InStock
37691-628
Erem® Cutting Swiss Tweezers, Cooper Tools
Forceps
Oblique heat cutting tweezers for cutting soft wire. Induction hardened cutting blades improves cutting capability and lifetime.
  • Made of tempered carbon steel
  • Anti-glare finish
This is a llong, anti-acid, medium curved tip tweezer and is perfect for use in micro-electronic applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty