Would you like to log in again?
NEEDLE TAPER TIP 18 GA PK1000
Supplier: APEX TOOL GROUP
Kahnetics* Tapered Tip Needle^ Taper reduces dripping from needle. Plastic material won't scratch delicate surfaces. Gauge: 18. I.D.: 0.86mm. Length: 38.1mm. Hub color: green.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
NEEDLE TAPER TIP 18 GA PK1000 37695-032 KDS18TNB Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty