Would you like to log in again?
NEEDLE THRD 20GA 1.5IN 1000/EA
Supplier: APEX TOOL GROUP
20 Gauge x 1.5in Threaded Hub Dispensing Needle^ Stainless steel plastic hub dispensing needles. All needles lock on to Luer Lok style syringes. Yellow hub. Inside diameter: 0.026in. Outside diameter: 0.035in. Length: 1.5in. 1000 needles per bag.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
NEEDLE THRD 20GA 1.5IN 1000/EA 37695-108 KDS20112BT Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty