Would you like to log in again?
Kahnetics® Tapered Needles, Cooper Tools
Supplier: Cooper Tools
Kahnetics®
KDS22TNP KDS18TNP KDS14TNP KDS16TNP
37695-028EA 67.78 USD InStock
37695-028 37695-148 37696-952 37696-998
Kahnetics® Tapered Needles, Cooper Tools
Fluid Adhesive Dispensers and Pistons
All-polyethylene dispensing tips
Tapered design reduces dripping. Polyethylene material won’t scratch delicate surfaces. Overall length: 38.1mm (11/2").
Ordering information: Tips are packaged 50 per pack.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty