Would you like to log in again?
SRYINGE AIR-OPERATED 30CC PK10
Supplier: APEX TOOL GROUP
Kahnetics* Air-Operated Syringe^ For micro-air dispensing. With barrel, rubber stopper, and Luer Lok* tip style. Unassembled and uncalibrated. Volume: 30mL.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SRYINGE AIR-OPERATED 30CC PK10 37695-252 A30LL Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty