Would you like to log in again?
TIP CON. .015 DIA .320L-MT150
Supplier: APEX TOOL GROUP
Conical Tip^ Diameter: 0.38mm. Reach: 8.1mm. For use with Weller* MicroTouch Plus* SMT/Thru-Hole Soldering Station (37698-506).
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TIP CON. .015 DIA .320L-MT150 37696-372 MT303 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty