Would you like to log in again?
Kahnetics® Stainless Steel Needles, Cooper Tools
Supplier: Cooper Tools
Kahnetics®
KDS16112P KDS1712P KDS2212P KDS181P KDS2312P KDS141P KDS1412P KDS221P KDS161P KDS201P KDS1512P KDS2512P KDS1912P
37695-138EA 40.21 USD InStock
37695-138 37695-168 37695-022 37695-010 37695-054 37696-990 37695-196 37695-140 37695-084 37696-986 37696-942 37696-974 37696-944
Kahnetics® Stainless Steel Needles, Cooper Tools
Syringe Needles
Blunt needles have plastic, color-coded hubs
All needles lock onto Luer-lock style syringes. Available in lengths from 12.7 to 38.1mm (1/2 to 11/2"). Disposable. Sold in packs of 50.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty