Would you like to log in again?
Weller® Soldering Irons, Cooper Tools, Apex
Supplier: Apex
Weller®
SL325
37698-712EA 182.73 USD InStock
37698-712
Weller® Soldering Irons, Cooper Tools, Apex
Soldering Supplies Soldering Irons
Modular soldering irons feature a heater for thread-on tips, a 1.5m (5') three-wire grounded cord, and a handle with acid- and flux-resistant cushioned grips.
Tips are designed to provide maximum heat transfer from heater to tip. They feature a copper core electroplated with iron to extend the lifetime of the tip. Nickel and chromium behind the working surface protect the shank from corrosion and solder creep.
Certifications: UL and C-UL listed.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty