Would you like to log in again?
HEATER 45W F/THREAD-IN TIPS
Supplier: APEX TOOL GROUP
Heater for 3.2mm Thread-In Tips^ 45W heater. Temperature: 482[degree]C. Use with Weller* Standard Series Modular Irons (37698-964, -962, -960).
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HEATER 45W F/THREAD-IN TIPS 37698-972 4035S Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty