Would you like to log in again?
Erem® Magic™ 'N' Series All-Purpose Diagonal Cutters, Cooper Tools
Supplier: Apex
Erem® Magic™
632N 522N
37699-416EA 219.99 USD InStock
37699-416 37699-368
Erem® Magic™ 'N' Series All-Purpose Diagonal Cutters, Cooper Tools
Cutters
These side cutters are including the features such as superior-grade tool steel construction, maximum opening stop, interlock screw joint system and ESD-safe cushion grips.
  • Oval head is the most widely used head shape
  • It is robust and size for size offers the highest cutting capacity
  • Internal springs for fatigue-and wobble-free operation
  • These cutters ensure high precision and extended life
The Cutting capacity varies based on the wire quality, these cutters are mainly suitable for cutting medium-hard wire, stainless steel wire, soft wire, copper and aluminum wires.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty