Would you like to log in again?
Erem® Oblique Angle Head Cutter, Flush Cut, Cooper Tools
Supplier: Apex
Erem®
503ETF
37699-458EA 618.84 USD InStock
37699-458
Erem® Oblique Angle Head Cutter, Flush Cut, Cooper Tools
Cutters
The angled head of this cutter provides for precise cuts at different working angles.
  • Head features a 30° angle
Carbide blades cut 18 AWG and smaller wires and mainly used for micro-electronic applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty