Would you like to log in again?
Xcelite® Precision Electronic Screwdriver Sets, Apex
Supplier: Apex
Xcelite®
XP600 XPTX600
37699-570EA 139.92 USD InStock
37699-570 37699-592
Xcelite® Precision Electronic Screwdriver Sets, Apex
Screwdrivers
For protection of ESD-sensitive items
The chrome-molybdenum-vanadium steel blades provide superior strength. Screwdrivers feature fast turning swivel caps with cavity to provide optimum control for precise centering, black oxide tips for longer wear life, no-roll handle design, and a storage case.
Set 37699-570 includes one each of No. 0 and No. 00 point Phillips® screwdrivers and slotted tip screwdrivers of the following sizes: 1.5, 2, 2.5, and 3 mm. TORX® set 37699-592 includes one each of No. 5 point and No. 6 point standard screwdrivers and four tamper-proof standard screwdrivers: No. 7, No. 8, No. 10, and No. 15 point.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty