Would you like to log in again?
Premium Back Support Belt, OK-1® Safety, OccuNomix
Supplier: OccuNomix
OK1000S M OK1000S L
39870-018CS 272.88 USD InStock
39870-018 39870-016
Premium Back Support Belt, OK-1® Safety, OccuNomix
Ergonomic Supports Back Supports
Featuring strong, power-knit nylon and elastic construction, these belts are designed for people who lift average weight loads or are involved in repetitive motion at work
  • Strong, power-knit nylon and elastic construction
  • For average weight loads or repetitive motion
  • Lumbar pad molds comfortably
  • Sturdy double-closure system overlaps twice for superior abdominal support
The lumbar pad molds comfortably into the lower lumbar region of the spine, while the sturdy double closure system overlaps twice for superior abdominal support.
Ordering information: All come with 2-way stretch, 4cm (11/2") wide suspenders, featuring breakaway swivel suspender clips that assure the suspenders detach only when you want them to.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty