Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Microtube Storage Boxes

Supplier: VWR
VWR®
R8300-P-VWR R8300-Y-VWR R8300-B-VWR R8300-G-VWR R8300-VWR R8300-A-VWR R8300-BLK-VWR R8300-O-VWR
40000-324CS 2412.89 USD InStock
40000-324 40000-326 40000-320 40000-332 40000-322 40000-330 40000-328 89084-934
VWR® Microtube Storage Boxes
Boxes Cryoboxes
These durable, polypropylene boxes maintain their integrity under harsh freezer conditions and will not absorb water or degrade like cardboard storage boxes. A three-point hinge and positive catch provide quick access to samples and keep them secure when box is closed. Boxes are available in seven colors. An alphanumeric grid on the lid aids in sample identification. Autoclavable to 121°C (249.8°F).

  • Racks for the chest freezers store up to nine boxes
  • Racks for the upright freezers store up to eight boxes
  • Each box holds one hundred 1.5 mL or 2.0 mL tubes
  • Aailable in seven colors (with clear lids)
  • Autoclavable to 121 °C (249.8 °F)
  • Temperature Range: –80 to 121 °C (–112 to 249.8 °F)

A three-point hinge and positive catch provide quick access to samples and keep them secure when box is closed. Alphanumeric indexing is printed inside the boxes which aids in sample identification.

Ordering information: Assorted boxes includes one box in each of the following colors: blue, green, orange, pink, and yellow.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty