Would you like to log in again?
FIBERPACK DRUM SQUARE 6GAL 6 GL
Supplier: GREIF BROTHERS CORP
Ro-Con* Transport Square Fibre Drum^ Size: 22.7L. Features rounded corners, convex sides, sturdy single wall construction, telescoping cover, and offset tape closure. For filling, handling, warehousing and/or shipping of dry chemicals products.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FIBERPACK DRUM SQUARE 6GAL 6 GL 40300-180 FRC8007 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty