Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VWR® Wide Mouth Laboratory Bottles, Amber HDPE
Supplier: VWR
VWR®
581340 581320 581330 581310 414004-118 414004-121 414004-120 414004-117 581300 414004-116 414004-119 581350
414004-117CS 360.7 USD InStock
414004-117 414004-116 89221-676 89221-678 414004-121 89221-672 414004-120 89221-674 89221-680 89221-670 414004-119 414004-118
VWR® Wide Mouth Laboratory Bottles, Amber HDPE
Bottles Plastic Bottles
Bottles are for general laboratory use, but are especially good for light-sensitive materials or photo- sensitive samples.
  • Ideal for light-sensitive materials or photosensitive samples
  • Leakproof
  • Easy-to-pour wide mouth design
Can be stored in freezer to −100 °C (−148 °F), and have excellent chemical resistance to alcohols, akalis, and both diluted and concetrated acids. Thick walls afford durability. Shrink ring at neck ensures smooth interior so that contents pour fluidly, while sealing ring and semi-buttressed threads in closure ensure leak prevention. Bottles are produced via the injection blow molding process for exact features and bottle stability.
Ordering information: Amber polypropylene caps included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty