Would you like to log in again?
VWR® Laboratory Bottles, HDPE
Supplier: VWR
VWR®
584220 583170 414004-158 414004-154 414004-155 583150 583120 414004-151 414004-152 584230 583140 583110 414004-159 584250 584200 583180 583100 584210 414004-156 414004-157 584240 414004-153 583190
414004-159CS 198.38 USD InStock
414004-159 414004-158 89221-590 414004-157 414004-156 414004-155 414004-154 89221-658 89221-588 89221-666 89221-668 414004-153 89221-584 89221-662 89221-596 414004-152 414004-151 89221-586 89221-664 89221-580 89221-582 89221-594 89221-660
VWR® Laboratory Bottles, HDPE
Bottles Plastic Bottles
Translucent, rigid bottles are for general laboratory use and for storing, shipping, and packing liquids.
  • Durable for long-term use
  • High-density polyethylene construction
  • Translucent siding
Can be stored in freezer to −100 °C (−148 °F). Excellent chemical resistance to most acids, bases, and alcohols. Can be used with most corrosives.
Delivery information: Polypropylene screw closure included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty