Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Laboratory Narrow Mouth Bottles, PP

Supplier: VWR
VWR®
414004-182 414004-178 414004-177 582150 414004-179 414004-183 582140 414004-181 414004-180 582100 582090 582120 582130 414004-176 582080
414004-179CS 166.53 USD InStock
414004-179 414004-178 414004-177 414004-176 89221-638 89221-632 89221-644 89221-634 89221-640 89221-630 414004-183 89221-642 414004-182 414004-181 414004-180
VWR® Laboratory Narrow Mouth Bottles, PP
Bottles Plastic Bottles
Specimen bottles are for general laboratory use, especially where sterilization is necessary.

  • Clear polypropylene construction
  • Screw enclosure included
  • Excellent chemical resistance
  • Can be stored in freezer to −40 °C (−40 °F)
  • Can be autoclaved with or without contents

Bottles are translucent and provide better clarity than polyethylene bottles

Note: Before autoclaving, set closure on top of container without engaging threads.

Delivery information: Polypropylene screw closure included.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty