Would you like to log in again?
VWR® Large Wide Mouth Bottles , PP
Supplier: VWR
VWR®
414004-187 414004-188
414004-188CS 232.02 USD InStock
414004-188 414004-187
VWR® Large Wide Mouth Bottles , PP
Bottles Plastic Bottles
Bottles are specifically designed for applications that require autoclaving of large volumes.
Note: Before autoclaving, set closure on top of bottle without engaging threads.
Ordering information: Bottle 414004-188 is supplied with a handle.
Delivery information: 100-415 white polypropylene screw cap included.
Caution: Bottles are not leakproof and are not recommended for shipping liquids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty