Would you like to log in again?
VWR® Wide Mouth Bottles, Rectangular, HDPE,
Supplier: VWR
VWR®
414004-193 414004-191 414004-192 414004-195 414004-194
414004-195CS 270.5 USD InStock
414004-195 414004-194 414004-193 414004-192 414004-191
VWR® Wide Mouth Bottles, Rectangular, HDPE,
Bottles Plastic Bottles
Space-saving, leakproof bottles are for sampling, storage, or shipment of dry chemicals or liquids.
  • Sturdy construction
  • Space-saving design
  • Leakproof
  • HDPE fabrication
Sturdy construction prevents bulging, translucent material allows for optical clarity, and wide mouth makes filling easy. Bottles have excellent chemical resistance to most acids, bases, and alcohols, and can be stored in freezer to −100 °C (−148 °F). Bottle 414004-195 features integrated handle for better grip, greater safety, and pouring convenience. Polypropylene screw caps included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty