Would you like to log in again?
VWR® Large Narrow Mouth Bottles, LDPE
Supplier: VWR
VWR®
414004-184 414004-204 414004-185
414004-204CS 150.95 USD InStock
414004-204 414004-185 414004-184
VWR® Large Narrow Mouth Bottles, LDPE
Bottles Plastic Bottles
Ideal for collecting, storing, or shipping a variety of contents from distilled water to corrosive chemicals and standards.
  • Leakproof
  • LDPE construction
They can be stored in a freezer to −100 °C (−148 °F), and are low in trace metals, with excellent chemical resistance to most acids, bases, and alcohols. Bottles have a built-in loop on the shoulder for identification tag attachment.
Delivery information: Polypropylene screw caps included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty