Would you like to log in again?
VWR® Wash Bottles
Supplier: Avantor
VWR®
560070 560060 560050 560080
414004-227CS 187.92 USD InStock
414004-227 414004-226 414004-225 414004-228
VWR® Wash Bottles
Bottles Wash Bottles
LDPE wash bottle features a polypropylene screw cap and stem that are molded into a one-piece unit for leakproof operation.
  • LDPE construction
  • Provided with a polypropylene screw cap
  • Leakproof operation
Dispensing tube produces a fine stream, but can be cut for a coarser flow.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty