Would you like to log in again?
VWR® Maximum Coveralls
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
414004-319 414004-314 414004-684 414004-313 414004-306 414004-685 414004-316 414004-318 414004-321 414004-312 414004-305 414004-310 414004-315 414004-309 414004-686 414004-317 414004-320 414004-683 414004-311 414004-308
414004-315CS 257.46 USD InStock
414004-315 414004-314 414004-313 414004-312 414004-686 414004-685 414004-311 414004-684 414004-321 414004-310 414004-320 414004-683 414004-309 414004-319 414004-308 414004-318 414004-306 414004-317 414004-316 414004-305
VWR® Maximum Coveralls
Disposable Garments
These are the cleanest and most durable coveralls available within the VWR® Protection line. Manufactured from a specially formulated breathable microporous fabric, these garments are distinguished by their exceptional comfort benefits and superior barrier protection capabilities.
  • Superior fluid and particulate barrier
  • Lowest level of particle shedding
  • Light chemical-splash resistance
  • Breathable to ensure user comfort
  • Soft, cloth-like fabric with maximum tear strength
  • Bonded seam option available
The coveralls provide disposable barrier protection against fluids, biologicals, particulates, and light chemical splash
Coveralls are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
All coveralls feature inset sleeves, front zipper, and elastic cuffs, ankles, and back. Also available with or without an attached elastic hood. Garments equipped with attached boots have a long-wear anti-skid sole. Serged seams are standard on all coveralls unless otherwise noted (bonded seams also available).
A high vapor transmission rate keeps the wearer comfortable, even throughout extended use. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty