Would you like to log in again?
VWR® Basic White SPP Coveralls
Supplier: VWR
VWR®
414004-338 414004-336 414004-341 414004-340 414004-337 414004-339 414004-335
414004-337CS 199.1 USD InStock
414004-337 414004-336 414004-335 414004-341 414004-340 414004-339 414004-338
VWR® Basic White SPP Coveralls
Disposable Garments
Coveralls are manufactured from a spunbonded polypropylene (SPP) fabric that is strong, lightweight, and breathable.
  • Economical, disposable first-line barrier
  • Maximum breathability
  • Low linting
They protect against dirt, grime, and certain dry particulates in nonhazardous environments
Garments feature inset sleeves, front zipper, tapered collar, and elastic cuffs, ankles, and back. No pockets. Serged seams. Also available in blue (30g/m²).
These general-purpose garments are ideal for less critical areas or pre-gowning entry rooms. Coveralls are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty